Anti-FASLG Hamster Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
106602
BLEG106602EA 370 EUR
BLEG106602
Anti-FASLG Hamster Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: FasL (CD178)
Clonality: Monoclonal
Clone:
Conjugation: PURE
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR