Anti-LY6A Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: D7]

Lieferant: BIOLEGEND INC
108129
BLEG108129EA 304 EUR
BLEG108129
Anti-LY6A Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: D7]
Antikörper
Type: Primary
Antigen: LY6A
Clonality: Monoclonal
Clone: D7
Conjugation: Brilliant Violet® 510
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR