Anti-LY6G Rat Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
108402
BLEG108402EA 161 EUR
BLEG108402
Anti-LY6G Rat Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: LY6G
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR