Anti-Mac-3 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]

Lieferant: BIOLEGEND INC
108504
BLEG108504EA 446 EUR
BLEG108504
Anti-Mac-3 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]
Antikörper
Type: Primary
Antigen: Mac-3
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR