Anti-NK1.1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
108731
BLEG108731EA 304 EUR
BLEG108731
Anti-NK1.1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: KLRB1C
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR