Anti-KLRB1C Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: PK136]

Lieferant: BIOLEGEND INC
108738
BLEG108738EA 641 EUR
BLEG108738
Anti-KLRB1C Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: PK136]
Antikörper
Type: Primary
Antigen: KLRB1C
Clonality: Monoclonal
Clone: PK136
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR