Anti-CD45.2 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 104]

Lieferant: BIOLEGEND INC
109831
BLEG109831EA 288 EUR
BLEG109831
Anti-CD45.2 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 104]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 104
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR