Anti-CD45.2 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 104]

Lieferant: BIOLEGEND INC
109833
BLEG109833EA 326 EUR
BLEG109833
Anti-CD45.2 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 104]
Antikörper
Type: Primary
Antigen: CD45.2
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR