Anti-I-Ak (Abk) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]

Lieferant: BIOLEGEND INC
109908
BLEG109908EA 454 EUR
BLEG109908
Anti-I-Ak (Abk) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]
Antikörper
Type: Primary
Antigen: I-Ak (Abk)
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR