Anti-ITGAX Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: N418]

Lieferant: BIOLEGEND INC
117329 117330
BLEG117329EA 277 EUR
BLEG117329 BLEG117330
Anti-ITGAX Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: N418]
Antikörper
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR