Anti-ITGAX Hamster Monoclonal Antibody (Brilliant Violet® 570) [clone: N418]

Lieferant: BIOLEGEND INC
117331
BLEG117331EA 320 EUR
BLEG117331
Anti-ITGAX Hamster Monoclonal Antibody (Brilliant Violet® 570) [clone: N418]
Antikörper
Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 570
Epitope:
Host: Hamster
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR