Anti-LY6C Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: HK1.4]

Lieferant: BIOLEGEND INC
128029
BLEG128029EA 320 EUR
BLEG128029
Anti-LY6C Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: HK1.4]
Antikörper
Type: Primary
Antigen: LY6C1
Clonality: Monoclonal
Clone: HK1.4
Conjugation: Brilliant Violet® 570
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR