Anti-ITGA2B Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: MWReg30]

Lieferant: BIOLEGEND INC
133911
BLEG133911EA 320 EUR
BLEG133911
Anti-ITGA2B Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: MWReg30]
Antikörper
Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: MWReg30
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR