Anti-IL7R Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: A7R34]

Lieferant: BIOLEGEND INC
135025
BLEG135025EA 355 EUR
BLEG135025
Anti-IL7R Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: A7R34]
Antikörper
Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: A7R34
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR