Anti-CD279 Antibody (Brilliant Violet 785) [clone: 29F.1A12]

Lieferant: BIOLEGEND INC
135225
BLEG135225EA 445 EUR
BLEG135225
Anti-CD279 Antibody (Brilliant Violet 785) [clone: 29F.1A12]
Antikörper
Type:
Antigen: CD279
Clonality:
Clone: 29F.1A12
Conjugation: Brilliant Violet 785
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR