Anti-MRC1 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: C068C2]

Lieferant: BIOLEGEND INC
141717
BLEG141717EA 320 EUR
BLEG141717
Anti-MRC1 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: C068C2]
Antikörper
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: C068C2
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR