Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]

Lieferant: BIOLEGEND INC
201509
BLEG201509EA 259 EUR
BLEG201509
Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
Antikörper
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR