Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: RPA-T4]

Lieferant: BIOLEGEND INC
300533
BLEG300533EA 326 EUR
BLEG300533
Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: RPA-T4]
Antikörper
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR