Anti-CD11A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]

Lieferant: BIOLEGEND INC
301212
BLEG301212EA 454 EUR
BLEG301212
Anti-CD11A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
Antikörper
Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Baboon, Bovine, Chimpanzee, Cynomolgus, Dog, Human, Rabbit, Rhesus, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR