Anti-ITGAM Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: ICRF44]

Lieferant: BIOLEGEND INC
301331
BLEG301331EA 326 EUR
BLEG301331
Anti-ITGAM Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: ICRF44]
Antikörper
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus, Common Marmoset, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR