Anti-CD14 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: M5E2]

Lieferant: BIOLEGEND INC
301837
BLEG301837EA 406 EUR
BLEG301837
Anti-CD14 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: M5E2]
Antikörper
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: M5E2
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chimpanzee, Cynomolgus, Rhesus, Capuchin Monkey, Cotton-topped Tamarin, Pigtailed Macaque, Common Marmoset, Squirrel Monkey, Bovine, Pig, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR