Anti-IGFR3 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 3G8]

Lieferant: BIOLEGEND INC
302035
BLEG302035EA 355 EUR
BLEG302035
Anti-IGFR3 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 3G8]
Antikörper
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus, Marmoset, Capuchin Monkey, Pigtailed Macaque, Sooty Mangabey, Squirrel Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR