Anti-FCGR3A Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: 3G8]

Lieferant: BIOLEGEND INC
302045
BLEG302045EA 436 EUR
BLEG302045
Anti-FCGR3A Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: 3G8]
Antikörper
Type: Primary
Antigen: FCGR3A
Clonality: Monoclonal
Clone: 3G8
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus, Marmoset, Capuchin Monkey, Pigtailed Macaque, Sooty Mangabey, Squirrel Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR