Anti-CD20 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 2H7]

Lieferant: BIOLEGEND INC
302331
BLEG302331EA 338 EUR
BLEG302331
Anti-CD20 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: 2H7]
Antikörper
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 2H7
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Capuchin Monkey, Squirrel Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR