Anti-IL2RA Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: BC96]

Lieferant: BIOLEGEND INC
302629 302630
BLEG302629EA 367 EUR
BLEG302629 BLEG302630
Anti-IL2RA Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: BC96]
Antikörper
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Rhesus, Pigtailed Macaque
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR