Anti-CD27 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: ]

Lieferant: BIOLEGEND INC
302835
BLEG302835EA 304 EUR
BLEG302835
Anti-CD27 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: ]
Antikörper
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR