Anti-CD38 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: ]

Lieferant: BIOLEGEND INC
303531
BLEG303531EA 326 EUR
BLEG303531
Anti-CD38 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: ]
Antikörper
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR