Anti-PTPRC Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: ]

Lieferant: BIOLEGEND INC
304021 304022 304029
BLEG304021EA 184 EUR
BLEG304021 BLEG304022 BLEG304029
Anti-PTPRC Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: ]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chimpanzee, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR