Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: HI30]

Lieferant: BIOLEGEND INC
304048
BLEG304048EA 658 EUR
BLEG304048
Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: HI30]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI30
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR