Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: HI100]

Lieferant: BIOLEGEND INC
304137
BLEG304137EA 338 EUR
BLEG304137
Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: HI100]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI100
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Chimpanzee
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR