Anti-CD45RO Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
304223
BLEG304223EA 304 EUR
BLEG304223
Anti-CD45RO Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL1
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Chimpanzee, Common Marmoset
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR