Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: UCHL1]

Lieferant: BIOLEGEND INC
304233
BLEG304233EA 363 EUR
BLEG304233
Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: UCHL1]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL1
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Chimpanzee, Common Marmoset
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR