Anti-CD80 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
305221
BLEG305221EA 338 EUR
BLEG305221
Anti-CD80 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 0
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR