Anti-CD86 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: IT2.2]

Lieferant: BIOLEGEND INC
305427
BLEG305427EA 380 EUR
BLEG305427
Anti-CD86 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: IT2.2]
Antikörper
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: IT2.2
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR