Anti-HLA-DR Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: L243]

Lieferant: BIOLEGEND INC
307637
BLEG307637EA 326 EUR
BLEG307637
Anti-HLA-DR Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: L243]
Antikörper
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: L243
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Pigtailed Macaque, Rhesus, Squirrel Monkey, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR