Anti-CD40L Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]

Lieferant: BIOLEGEND INC
310823
BLEG310823EA 338 EUR
BLEG310823
Anti-CD40L Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ]
Antikörper
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 24-31
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR