Anti-CD24 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ML5]

Lieferant: BIOLEGEND INC
311121 311122
BLEG311121EA 303 EUR
BLEG311121 BLEG311122
Anti-CD24 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: ML5]
Antikörper
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: ML5
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chimpanzee, Baboon
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR