Anti-MME Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: HI10a]

Lieferant: BIOLEGEND INC
312222
BLEG312222EA 667 EUR
BLEG312222
Anti-MME Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: HI10a]
Antikörper
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: HI10a
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus, Capuchin monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR