Anti-ICOS Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: C398.4A]

Lieferant: BIOLEGEND INC
313524
BLEG313524EA 667 EUR
BLEG313524
Anti-ICOS Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: C398.4A]
Antikörper
Type: Primary
Antigen: ICOS
Clonality: Monoclonal
Clone: C398.4A
Conjugation: Brilliant Violet® 421
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rhesus, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR