Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]

Lieferant: BIOLEGEND INC
317307 317308
BLEG317307EA 35.1 EUR
BLEG317307 BLEG317308
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]
Antikörper
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 0
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR