Anti-CD3E Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: OKT3]

Lieferant: BIOLEGEND INC
317329
BLEG317329EA 338 EUR
BLEG317329
Anti-CD3E Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: OKT3]
Antikörper
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: OKT3
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR