Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: OKT4]

Lieferant: BIOLEGEND INC
317433 317434
BLEG317433EA 277 EUR
BLEG317433 BLEG317434
Anti-CD4 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: OKT4]
Antikörper
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR