Anti-NCAM1 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: HCD56]

Lieferant: BIOLEGEND INC
318335
BLEG318335EA 387 EUR
BLEG318335
Anti-NCAM1 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: HCD56]
Antikörper
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: HCD56
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol NCAM1
Environmentally Preferable
Antigen-Name Neural Cell Adhesion Molecule 1
Antigen-Synonyme NCAM|MSK39|CD56
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Brilliant Violet® 711
Klon HCD56
Reaktivität Baboon,Macaque,Chimpanzee,Human
Wirt Mouse
Gen-ID 4684
Isotyp IgG1 kappa
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Reinigung The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 711® under optimal conditions. The solution is free of unconjugated Brilliant Violet 711® and unconjugated antibody.
Lagertemperatur The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Transporttemperatur Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR