Multiple Isotype Control (FITC (Fluorescein Isothiocyanate)/PE (Phycoerythrin))

Lieferant: BIOLEGEND INC
319801
BLEG319801EA 304 EUR
BLEG319801
Multiple Isotype Control (FITC (Fluorescein Isothiocyanate)/PE (Phycoerythrin))
Antikörper
Conjugations: FITC (Fluorescein Isothiocyanate), PE (Phycoerythrin)

Type: Control
Antigen: IgG1/IgG2a
Clonality: Monoclonal
Clone:
Conjugation: Various
Epitope:
Host:
Isotype: Multiple
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR