Anti-CD314 Mouse Monoclonal Antibody (Biotin) [clone: ]

Lieferant: BIOLEGEND INC
320804
BLEG320804EA 312 EUR
BLEG320804
Anti-CD314 Mouse Monoclonal Antibody (Biotin) [clone: ]
Antikörper
Type: Primary
Antigen: CD314
Clonality: Monoclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR