Anti-CD15 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: ]

Lieferant: BIOLEGEND INC
323027
BLEG323027EA 288 EUR
BLEG323027
Anti-CD15 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: ]
Antikörper
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR