Anti-HLA-DR Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: ]

Lieferant: BIOLEGEND INC
327012
BLEG327012EA 387 EUR
BLEG327012
Anti-HLA-DR Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: ]
Antikörper
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rhesus monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR