Anti-THY1 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 5E10]

Lieferant: BIOLEGEND INC
328128
BLEG328128EA 667 EUR
BLEG328128
Anti-THY1 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 5E10]
Antikörper
Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: 5E10
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR