Anti-CD274 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: 29E.2A3]

Lieferant: BIOLEGEND INC
329722
BLEG329722EA 658 EUR
BLEG329722
Anti-CD274 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: 29E.2A3]
Antikörper
Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 29E.2A3
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, African Green, Chimpanzee, Common Marmoset, Squirrel Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR