Anti-PD1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: EH12.2H7]

Lieferant: BIOLEGEND INC
329920 329919
BLEG329920EA 624 EUR
BLEG329920 BLEG329919
Anti-PD1 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: EH12.2H7]
Antikörper
Type: Primary
Antigen: PDCD1
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chimpanzee, Common Marmoset, Rhesus, Squirrel Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR